missing translation for 'onlineSavingsMsg'
Learn More

Glut5 Antibody (OTI9F3), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP1-47980

Product Code. 18242781

  • £487.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Glut5 Monoclonal specifically detects Glut5 in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Glut5
Monoclonal
1 mg/mL
Western Blot 1:500-1000, Flow Cytometry 1:100, Immunohistochemistry 1:50, Immunoprecipitation 2ug/500ul, Immunohistochemistry-Paraffin 1:50
SLC2A5
Full length human recombinant protein of human SLC2A5 (NP_003030) produced in HEK293T cell.
Affinity Purified
RUO
6518
Human, Monkey
IgG1
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
OTI9F3
Unconjugated
Fructose transporter, Glucose transporter type 5, small intestine, GLUT-5, GLUT5glucose transporter-like protein 5, solute carrier family 2 (facilitated glucose transporter), member 5, solute carrier family 2 (facilitated glucose/fructose transporter), member 5, solute carrier family 2, facilitated glucose transporter member 5
Mouse
54.8 kDa
0.1 mL
Primary
This antibody is specific for Homo sapiens solute carrier family 2 (facilitated glucose/fructose transporter), member 5 (SLC2A5), transcript variant 1.
Store at -20°C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only