missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glut5 Antibody (195205) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1349JF585
This item is not returnable.
View return policy
Description
Glut5 Monoclonal antibody specifically detects Glut5 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,CyTOF
Specifications
| Glut5 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| Fructose transporter, Glucose transporter type 5, small intestine, GLUT-5, GLUT5glucose transporter-like protein 5, solute carrier family 2 (facilitated glucose transporter), member 5, solute carrier family 2 (facilitated glucose/fructose transporter), member 5, solute carrier family 2, facilitated glucose transporter member 5 | |
| NS0 mouse myeloma cell line transfected with human Glut5, Met1-Gln501, Accession # P22732 | |
| RUO | |
| 6518 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 195205 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.2 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction