missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glut4 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-49840
This item is not returnable.
View return policy
Description
Glut4 Polyclonal specifically detects Glut4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| Glut4 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| GLUT-4, GLUT4Glucose transporter type 4, insulin-responsive, insulin-responsive glucose transporter type 4, solute carrier family 2 (facilitated glucose transporter), member 4, solute carrier family 2, facilitated glucose transporter member 4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is specific to the C Terminus. Expected from sequence similarity: Human, Cow, Pig. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3-1 ug/ml, Immunohistochemistry-Paraffin 3.75 ug/ml Peptide ELISA Detection limit 1:16000 | |
| P14672 | |
| SLC2A4 | |
| Peptide with sequence C-TELEYLGPDEND corresponding to C-Terminus according to NP_001033.1. | |
| 0.1 mg | |
| Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones | |
| 6517 | |
| Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction