missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GLUT3 Recombinant Rabbit Monoclonal Antibody (K01_1R71)

Product Code. 30828216
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828216 50 μL
30829386 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828216 Supplier Invitrogen™ Supplier No. MA565062
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

GLUT3 Recombinant Monoclonal Antibody for Western Blot

Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF1 induced signaling. GLUT3 is detectable in a few normal cell type spermatids in testis with active spermatogenesis, placental trophoblast membranes, and neurons in brain. GLUT3 staining is also detectable in human cancers including those of the ovary, lung, and testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLUT3
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1R71
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene SLC2A3
Gene Accession No. P11169, P32037, Q07647
Gene Alias AA408729; AL023014; AL024341; AU040424; C78366; FLJ90380; glucose transporter 3; glucose transporter type 3, brain; Glut3; Glut-3; Slc2a3; solute carrier family 2 (facilitated glucose transporter), member 3; Solute carrier family 2 A3 (neuron glucose transporter); solute carrier family 2 member 3; solute carrier family 2, facilitated glucose transporter member 3; solute carrier family 2, member 2; solute carrier family 2, member 3
Gene Symbols SLC2A3
Host Species Rabbit
Immunogen A synthetic peptide of human Glucose Transporter GLUT3.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20527, 25551, 6515
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.