missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLUT2 Monoclonal Antibody (205115)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA523807
This item is not returnable.
View return policy
Description
This antibody recognizes mouse Glut2-transfected NS0 cells but not NS0 control transfectants. This antibody also detects Glut2 on mouse insulinoma beta TC-6 cells. Reconstitute at 0.5 mg/mL in sterile PBS.
Glucose transporter 2 isoform is an integral plasma membrane glycoprotein of the liver, islet beta cells, intestine, and kidney epithelium. It mediates facilitated bidirectional glucose transport. Because of its low affinity for glucose, it has been suggested as a glucose sensor. Mutations in the gene that encode this protein cause Fanconi-Bickel syndrome.
Specifications
| GLUT2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P12336, P14246 | |
| Slc2a2 | |
| CHO Chinese hamster ovary cell line transfected with mouse Glut2 Met1-Val523. | |
| 200 μg | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 205115 | |
| Unconjugated | |
| Slc2a2 | |
| AI266973; glucose transporter 2; glucose transporter type 2, liver; GLUT2; GLUT-2; GTT2; liver-type glucose transporter; Slc2a2; solute carrier family 2 (facilitated glucose transporter), member 2; solute carrier family 2 member 2; solute carrier family 2, facilitated glucose transporter member 2 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 20526, 25351 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction