missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glut2 Antibody (205115) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1440JF635
This item is not returnable.
View return policy
Description
Glut2 Monoclonal antibody specifically detects Glut2 in Mouse samples. It is validated for Flow Cytometry,Immunocytochemistry,CyTOF
Specifications
| Glut2 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| GLUT-2, GLUT2 Glucose transporter type 2, liver, SLC2A2, solute carrier family 2 (facilitated glucose transporter), member 2, solute carrier family 2, facilitated glucose transporter member 2 | |
| CHO Chinese hamster ovary cell line transfected with mouse Glut2, Met1-Val523, Accession # P14246 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 205115 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.2 mL | |
| Lipid and Metabolism, Stem Cell Signaling Pathway | |
| 6514 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction