missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLUT14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121934
This item is not returnable.
View return policy
Description
SLC2A14 (Solute Carrier Family 2 Member 14) is a Protein Coding gene. Diseases associated with SLC2A14 include Hypoascorbemia and Amyotrophic Lateral Sclerosis 1. Among its related pathways are Metabolism and Transport of glucose and other sugars, bile salts and organic acids, metal ions and amine compounds. Gene Ontology (GO) annotations related to this gene include transmembrane transporter activity and glucose transmembrane transporter activity. An important paralog of this gene is SLC2A3.
Specifications
| GLUT14 | |
| Polyclonal | |
| Unconjugated | |
| SLC2A14 | |
| GLUT-14; GTR14 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 144195 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8TDB8 | |
| SLC2A14 | |
| Recombinant GLUT14 (Met1-Val105) expressed in E.coli. | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction