missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLUT10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583529
This item is not returnable.
View return policy
Description
Immunogen sequence: LPAGTDETAT HKDLIPLQGG EAPKLGPGRP RYSFLDLFRA RDNMRG Highest antigen sequence indentity to the following orthologs: Mouse - 63%, Rat - 61%.
Non-insulin-dependent diabetes mellitus (NIDDM) is a multifactoral disease with both environmental and genetics causes. Genome-wide screening procedures have identified several susceptibility loci for NIDDM within the human genome. A putative sugar transporter that has been localized to human chromosome 20q12-q13.1, one of the genomic loci associated with NIDDM. Because of the strong resemblance of this novel protein to members of the mammalian facilitative glucose transporter family (GLUT), the protein is known as GLUT10 (HGMW-approved gene symbol SLC2A10). Data suggests that GLUT10 an excellent candidate for a susceptibility gene involved in NIDDM.
Specifications
| GLUT10 | |
| Polyclonal | |
| Unconjugated | |
| SLC2A10 | |
| AA450473; ATS; glucose transporter type 10; Glut10; GLUT-10; GLUT11; MGC118830; MGC118833; Slc2a10; SLC2A11-a; SLC2A11-c; solute carrier family 2 (facilitated glucose transporter), member 10; solute carrier family 2 member 10; solute carrier family 2, facilitated glucose transporter member 10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 81031 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95528 | |
| SLC2A10 | |
| Recombinant protein corresponding to Human SLC2A10. Recombinant protein control fragment (Product #RP-99462). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction