missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLUT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21829-1-AP-20UL
This item is not returnable.
View return policy
Description
21829-1-AP antibody can also detect the 25 kDa degradation protein in SDS-PAGE.
Facilitative glucose transporter. This isoform may be responsible for constitutive or basal glucose uptake. Has a very broad substrate specificity; can transport a wide range of aldoses including both pentoses and hexoses.Specifications
| GLUT1 | |
| Polyclonal | |
| Unconjugated | |
| SLC2A1 | |
| DYT17, DYT18, GLUT, GLUT 1, GLUT1, HepG2 glucose transporter, PED, SLC2A1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20525, 24778, 6513 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P11166, P17809 | |
| SLC2A1 | |
| GLUT1 Fusion Protein Ag16282 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction