missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glut1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£232.00 - £419.00
Specifications
| Antigen | Glut1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18243640
|
Novus Biologicals
NBP2-54745 |
100 μL |
£419.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18642317
|
Novus Biologicals
NBP2-54745-25ul |
25 μL |
£232.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glut1 Polyclonal specifically detects Glut1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Glut1 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cardiovascular Biology, Cellular Markers, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroscience, Plasma Membrane Markers, Signal Transduction | |
| PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
| 6513 | |
| This Glut1 antibody was developed against a recombinant protein corresponding to amino acids: RNEENRAKSVLKKLRGTADVTHDLQE | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Choreoathetosis/Spasticity, Episodic (Paroxysmal Choreoathetosis), CSE, DYT17, DYT18, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, GLUT-1, GLUT1DS, HepG2 glucose transporter, HTLVR, Human T-Cell Leukemia Virus (I and II) Receptor, MGC141895, MGC141896, PED, solute carrier family 2 (facilitated glucose transporter), member 1, Solute Carrier Family 2 Member 1, solute carrier family 2, facilitated glucose transporter member 1 | |
| SLC2A1 | |
| IgG | |
| Affinity Purified | |
| 54.1 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title