missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glut1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 23 publications
Brand: Novus Biologicals NB300-666
This item is not returnable.
View return policy
Description
Glut1 Polyclonal specifically detects Glut1 in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Glut1 | |
| Polyclonal | |
| Unconjugated | |
| Tris/Glycine buffer (pH 7.4 - 7.8), HEPES, 0.5% BSA, 30% glycerol with 0.02% Sodium Azide | |
| Choreoathetosis/Spasticity, Episodic (Paroxysmal Choreoathetosis), CSE, DYT17, DYT18, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, GLUT-1, GLUT1DS, HepG2 glucose transporter, HTLVR, Human T-Cell Leukemia Virus (I and II) Receptor, MGC141895, MGC141896, PED, solute carrier family 2 (facilitated glucose transporter), member 1, Solute Carrier Family 2 Member 1, solute carrier family 2, facilitated glucose transporter member 1 | |
| Rabbit | |
| 54.1 kDa | |
| 0.1 mg | |
| Cancer, Cardiovascular Biology, Cellular Markers, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroscience, Plasma Membrane Markers, Signal Transduction | |
| 6513 | |
| Human, Mouse, Rat, Zebrafish | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.66mg/mL | |
| Western Blot 1:5000, ELISA 1:12000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| P11166 | |
| SLC2A1 | |
| This Glut1 antibody is made against a synthetic peptide conjugated to KLH, corresponding to amino acids 478-492 of Human Glucose Transporter GLUT1. Peptide was covalently modified. Sequence: PEELFHPLGADSQV | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Glucose Transporter GLUT1 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction