missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glut1 Antibody (GLUT1/2475), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-75786F
This item is not returnable.
View return policy
Description
Glut1 Monoclonal specifically detects Glut1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Glut1 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| SLC2A1 | |
| Recombinant fragment of human Glut1 protein (around aa 203-305) (exact sequence is proprietary) (Uniprot: P11166) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| IgG2b κ |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| GLUT1/2475 | |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Choreoathetosis/Spasticity, Episodic (Paroxysmal Choreoathetosis), CSE, DYT17, DYT18, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, GLUT-1, GLUT1DS, HepG2 glucose transporter, HTLVR, Human T-Cell Leukemia Virus (I and II) Receptor, MGC141895, MGC141896, PED, solute carrier family 2 (facilitated glucose transporter), member 1, Solute Carrier Family 2 Member 1, solute carrier family 2, facilitated glucose transporter member 1 | |
| Mouse | |
| 54.1 kDa | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Markers, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroscience, Plasma Membrane Markers, Signal Transduction | |
| Recognizes a protein of 54.1 kDa, which is identified as GLUT-1. Glucose transporters are integral membrane glycoproteins involved in transporting glucose into most cells. There are many types of glucose transport carrier proteins, designated as Glut-1 to Glut-12. Glut-1 is a major glucose transporter in the mammalian blood-brain barrier. It is expressed in high density on the membranes of human erythrocytes and the brain capillaries that comprise the blood-brain barrier. Glut-1 is expressed at variable levels in many human tissues. Overexpression of Glut-1 has been linked to tumor progression or poor survival of patients with carcinomas of the colon, breast, cervical, lung, bladder and mesothelioma. Glut-1 is a sensitive and specific marker for the differentiation of malignant mesothelioma (positive) from reactive mesothelium (negative). | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction