missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GluR7 Polyclonal Antibody

Description
A suggested positive control is human brain tissue lysate. PA5-20450 can be used with blocking peptide PEP-0567.
Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. This gene product belongs to the kainate family of glutamate receptors, which are composed of four subunits and function as ligand-activated ion channels. It is not certain if the subunit encoded by this gene is subject to RNA editing as the other 2 family members (GRIK1 and GRIK2). A Ser310Ala polymorphism has been associated with schizophrenia, and there are conflicting reports of its association with the pathogenesis of delirium tremens in alcoholics.
Specifications
Specifications
| Antigen | GluR7 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | GRIK3 |
| Gene Accession No. | B1AS29, P42264, Q13003 |
| Gene Alias | 9630027E11; dJ1090M5.1 (glutamate receptor, ionotropic, kainate 3 (GLUR7)); EAA5; Excitatory amino acid receptor 5; GLR7; GluK3; GLUR7; Glur-7; GluR7-3; GluR7a; glutamate ionotropic receptor kainate type subunit 3; glutamate receptor; glutamate receptor 7; Glutamate receptor ionotropic, kainate 3; glutamate receptor subunit kainate subtype; glutamate receptor, ionotropic, kainate 3; Grik3 |
| Gene Symbols | GRIK3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?