missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GluR6 Recombinant Rabbit Monoclonal Antibody (24GB3695)

Rabbit Recombinant Monoclonal Antibody
£126.00 - £434.00
Specifications
| Antigen | GluR6 |
|---|---|
| Clone | 24GB3695 |
| Content And Storage | -20°C |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275021
![]() |
Invitrogen™
MA557033 |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276274
|
Invitrogen™
MA557034 |
20 μL |
£126.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GluR6 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Glutamate receptors are the primary excitatory neurotransmitter receptors in the mammalian brain and are activated during various normal neurophysiological processes. The kainate family of glutamate receptors, which are composed of four subunits and function as ligand-activated ion channels, includes this gene product. The subunit encoded by this gene is edited at multiple sites within the first and second transmembrane domains, which is thought to alter the structure and function of the receptor complex. Furthermore, different isoforms of this gene have been described due to alternatively spliced transcript variants. Mutations in this gene have been linked to autosomal recessive cognitive disability. GRIK2-associated diseases include Neurodevelopmental Disorder with Impaired Language and Ataxia and with or without Seizures and Intellectual Developmental Disorder, Autosomal Recessive 6.Specifications
| GluR6 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| GRIK2 | |
| AW124492; bA487F5.1; EAA4; Excitatory amino acid receptor 4; GLR 6; GLR6; GluK2; GLUK6; GLUR 6; gluR beta-2; Glur6; Glur-6; Glurbeta2; glutamate ionotropic receptor kainate type subunit 2; glutamate receptor; glutamate receptor 6; Glutamate receptor beta-2; glutamate receptor form A; glutamate receptor form B; glutamate receptor form C; glutamate receptor form D; glutamate receptor form E; glutamate receptor ionotropic, kainate 2; glutamate receptor, ionotropic kainate 2; glutamate receptor, ionotropic, kainate 2; glutamate receptor, ionotropic, kainate 2 (beta 2); glutamate receptor, ionotropic, kainate 5; GRIK 2; Grik2; GRIK2 protein; grik2alpha; grik2l; MRT6 | |
| GRIK2 | |
| Primary | |
| Affinity Chromatography |
| 24GB3695 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P39087, P42260, Q13002 | |
| 14806, 2898, 54257 | |
| A synthesized peptide derived from human GRIK2 (aa 100-150). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title