missing translation for 'onlineSavingsMsg'
Learn More

GluR5/GRIK1 Antibody, Novus Biologicals™

Product Code. 18148419 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18148419 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18148419 Supplier Novus Biologicals Supplier No. NB10068165

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

GluR5/GRIK1 Polyclonal specifically detects GluR5/GRIK1 in Human samples. It is validated for Western Blot, Peptide ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluR5/GRIK1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_000821.1
Gene Alias EEA3, GLR5, GluR5, GluR-5, glutamate receptor, ionotropic, kainate 1, ionotropic kainate 1
Gene Symbols GRIK1
Host Species Goat
Immunogen Peptide with sequence QCKQTHPTNSTS corresponding to internal region (near C-Terminus) according to NP_783300.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 2897
Test Specificity This antibody is expected to recognize both reported isoforms according to NP_000821.1 and NP_783300.1.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.