missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GluR5/GRIK1 Polyclonal antibody specifically detects GluR5/GRIK1 in Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | GluR5/GRIK1 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA 1:10000 |
| Formulation | Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide |
| Gene Alias | EEA3, GLR5, GluR5, GluR-5, glutamate receptor, ionotropic, kainate 1, ionotropic kainate 1 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to unique amino acid region on GluR5. |
| Purification Method | Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?