missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GluR2 Recombinant Rabbit Monoclonal Antibody (9R1X2)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535096
This item is not returnable.
View return policy
Description
Immunogen sequence: YLYDSDRGLS TLQAVLDSAA EKKWQVTAIN VGNINNDKKD EMYRSLFQDL ELKKERRVIL DCERDKVNDI VDQVITIGKH VKGYHYIIAN LGFTDGDLLK I.
The ion channels activated by glutamate are typically divided into two classes. Those that are sensitive to N-methyl-D-aspartate (NMDA) are designated NMDA receptors (NMDAR) while those activated by alpha-amino-3-hydroxy-5 -methyl-4-isoxalone propionic acid (AMPA) are known as AMPA receptors (AMPAR). The AMPAR are comprised of four distinct Glutamate Receptor Subunits designated (GluR1-4) and they play key roles in virtually all excitatory neurotransmission in the brain. The GluR2 subunit is thought to play a key role in forms of synaptic plasticity such as LTD.
Specifications
| GluR2 | |
| Recombinant Monoclonal | |
| 1.2 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P19491, P23819, P42262 | |
| Gria2 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 9R1X2 | |
| Unconjugated | |
| Gria2 | |
| AMPA selective glutamate receptor; AMPA-selective glutamate receptor 2; GluA2; Glur2; GluR-2; GLURB; GluR-B; GluR-K2; glutamate ionotropic receptor AMPA type subunit 2; glutamate receptor; glutamate receptor 2; glutamate receptor B; glutamate receptor B flip isoform; glutamate receptor ionotropic, AMPA 2; glutamate receptor, ionotropic, AMPA 2; glutamate receptor, ionotropic, AMPA2 (alpha 2); GRIA2; HBGR2; OTTHUMP00000165324; OTTHUMP00000220942; OTTHUMP00000220943 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14800, 2891, 29627 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction