missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GluR2 Antibody (7G6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£173.00 - £443.00
Specifications
| Antigen | GluR2 |
|---|---|
| Clone | 7G6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18637305
|
Novus Biologicals
NBP2-37475-0.025ml |
0.025 mL |
£173.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18098505
|
Novus Biologicals
NBP2-37475 |
0.1 mL |
£443.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GluR2 Monoclonal specifically detects GluR2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| GluR2 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| Unconjugated | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 2891 | |
| Purified recombinant fragment of human GRIA2 (AA: 652-807) expressed in E. Coli. | |
| Primary | |
| 98.8 kDa |
| 7G6 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| AMPA 2, glutamate receptor, ionotropic, AMPA 2 | |
| GRIA2 | |
| IgG1 | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title