missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GluR2 Antibody (7A7A3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£158.00 - £435.00
Specifications
| Antigen | GluR2 |
|---|---|
| Clone | 7A7A3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18260255
|
Novus Biologicals
NBP2-61775 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18664727
|
Novus Biologicals
NBP2-61775-0.025ml |
0.025 mL |
£158.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GluR2 Monoclonal specifically detects GluR2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| GluR2 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| AMPA 2, glutamate receptor, ionotropic, AMPA 2 | |
| GRIA2 | |
| IgG2b | |
| Protein G purified |
| 7A7A3 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Purified | |
| Apoptosis, Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Tumor Suppressors, Vision | |
| 2891 | |
| Purified recombinant fragment of human GRIA2 (AA: 35-175) expressed in E. Coli. | |
| Primary | |
| 99 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title