missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GluR2/3 Antibody (JE33-28), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32478
This item is not returnable.
View return policy
Description
GluR2/3 Monoclonal antibody specifically detects GluR2/3 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| GluR2/3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| AMPA 2, glutamate receptor, ionotropic, AMPA 2 | |
| Synthetic peptide within Human GRIA2 aa 834-883/ 883. (Uniprot: P42262) | |
| 100 μL | |
| Apoptosis, Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Tumor Suppressors, Vision | |
| 2891 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE33-28 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction