missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GluR1 Antibody (S355-1), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22399AF350
This item is not returnable.
View return policy
Description
GluR1 Monoclonal antibody specifically detects GluR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| GluR1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| S355-1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| AMPA 1, AMPA-selective glutamate receptor 1, GluA1, GLUH1, GluR-1, GLUR1gluR-A, GluR-A, GLURA, GluR-K1, glutamate receptor 1, Glutamate receptor ionotropic, AMPA 1, glutamate receptor, ionotropic, AMPA 1, HBGR1, MGC133252 | |
| Fusion protein amino acids 1-389 (extracellular N-terminus) of rat GluA1/GluR1 | |
| 0.1 mL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Vision | |
| 2890 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction