missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLUD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14299-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a subunit of glutamate receptor channels. These channels mediate most of the fast excitatory synaptic transmission in the central nervous system and play key roles in synaptic plasticity.Specifications
| GLUD1 | |
| Polyclonal | |
| Unconjugated | |
| GLUD1 | |
| GDH, GDH 1, GDH1, GLUD, GLUD1, glutamate dehydrogenase 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14661, 24399, 2746 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P00367, P10860, P26443 | |
| GLUD1 | |
| GLUD1 Fusion Protein Ag5694 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction