missing translation for 'onlineSavingsMsg'
Learn More

GLUD1/GLUD2 Antibody (848209) [DyLight 550], Novus Biologicals™

Product Code. 30646060 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30646060 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30646060 Supplier Novus Biologicals Supplier No. FAB8027L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GLUD1/GLUD2 Monoclonal antibody specifically detects GLUD1/GLUD2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLUD1/GLUD2
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 848209
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias epididymis secretory sperm binding protein, epididymis tissue sperm binding protein Li 18mP, GDH, GDH1, GLUD, glutamate dehydrogenase (NAD(P)+), glutamate dehydrogenase 1, mitochondrial, hGDH1
Host Species Mouse
Immunogen E. coli-derived recombinant human GLUD1/GLUD2, Ser54-Thr558, Accession # P49448
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2746
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.