missing translation for 'onlineSavingsMsg'
Learn More

Glucuronosyltransferase 1A1/UGT1A1 Antibody [HRP], Novus Biologicals™

Product Code. 30736640 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30736640 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736640 Supplier Novus Biologicals Supplier No. AF6490H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Glucuronosyltransferase 1A1/UGT1A1 Polyclonal antibody specifically detects Glucuronosyltransferase 1A1/UGT1A1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glucuronosyltransferase 1A1/UGT1A1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias bilirubin UDP-glucuronosyltransferase 1-1, bilirubin UDP-glucuronosyltransferase isozyme 1, Bilirubin-specific UDPGT isozyme 1, GNT1EC 2.4.1.17, HUG-BR1, UDP glucuronosyltransferase 1 family, polypeptide A1, UDP glycosyltransferase 1 family, polypeptide A1, UDP-glucuronosyltransferase 1-1, UDP-glucuronosyltransferase 1-A, UDP-glucuronosyltransferase 1A1, UDPGT, UGT1*1, UGT1.1, UGT1-01, UGT-1A, UGT1AUDPGT 1-1, UGT1BILIQTL1
Host Species Goat
Immunogen E. coli-derived recombinant human Glucuronosyltransferase 1A1/UGT1A1, Leu60-Thr168, Accession # P22309
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54658
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.