missing translation for 'onlineSavingsMsg'
Learn More

Glucuronosyltransferase 1A1/UGT1A1 Antibody (856754) [DyLight 350], Novus Biologicals™

Product Code. 30643211 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30643211 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30643211 Supplier Novus Biologicals Supplier No. FAB6490D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Glucuronosyltransferase 1A1/UGT1A1 Monoclonal antibody specifically detects Glucuronosyltransferase 1A1/UGT1A1 in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glucuronosyltransferase 1A1/UGT1A1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 856754
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias bilirubin UDP-glucuronosyltransferase 1-1, bilirubin UDP-glucuronosyltransferase isozyme 1, Bilirubin-specific UDPGT isozyme 1, GNT1EC 2.4.1.17, HUG-BR1, UDP glucuronosyltransferase 1 family, polypeptide A1, UDP glycosyltransferase 1 family, polypeptide A1, UDP-glucuronosyltransferase 1-1, UDP-glucuronosyltransferase 1-A, UDP-glucuronosyltransferase 1A1, UDPGT, UGT1*1, UGT1.1, UGT1-01, UGT-1A, UGT1AUDPGT 1-1, UGT1BILIQTL1
Host Species Mouse
Immunogen E. coli-derived recombinant human Glucuronosyltransferase 1A1/UGT1A1, Leu60-Thr168, Accession # P22309
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54658
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.