missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucuronic Acid Epimerase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-31872
This item is not returnable.
View return policy
Description
Glucuronic Acid Epimerase Polyclonal specifically detects Glucuronic Acid Epimerase in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Glucuronic Acid Epimerase | |
| Polyclonal | |
| Flow Cytometry, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| O94923 | |
| GLCE | |
| This antibody was developed against a recombinant protein corresponding to amino acids: ETSEGVSLQLGNTKDFIISFDLKFLTNGSVSVVLETTEKNQLFTIHYVSNAQLIAFKERDIYYGIGPRTSWSTVTRDLVTDLRKGVGLSNTKAVKPTKIMPKKVVRLI | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| D-glucuronyl C5-epimerase, EC 5.1.3, EC 5.1.3.-, glucuronic acid epimerase, glucuronyl C5-epimerase, Heparin/heparan sulfate:glucuronic acid C5-epimerase, HSEPIheparin/heparan sulfate-glucuronic acid C5-epimerase, KIAA0836heparan sulfate epimerase, UDP-glucuronic acid epimerase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 26035 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction