missing translation for 'onlineSavingsMsg'
Learn More

Glucosyl-C18-sphingosine, TRC

Product Code. 30012775
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012775 2.5 mg 2.5mg
30009387 10 mg 10mg
30006998 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012775 Supplier Toronto Research Chemicals Supplier No. TRCG5968402.5MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52050-17-6
Molecular Formula C24H47NO7
Molecular Weight (g/mol) 461.63
Synonym Glucosylsphingosine, 2-Amino-3-hydroxy-4-octadecenyl beta-D-Glucopyranoside, (2S,3R,4E)-2-Amino-3-hydroxy-4-octadecenyl beta-D-Glucopyranoside
IUPAC Name (2R,3R,4S,5S,6R)-2-[(E,2S,3R)-2-amino-3-hydroxyoctadec-4-enoxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCCCCCCCCCCCC\C=C\C(O)C(N)CO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C24H47NO7/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(27)18(25)17-31-24-23(30)22(29)21(28)20(16-26)32-24/h14-15,18-24,26-30H,2-13,16-17,25H2,1H3/b15-14+/t18-,19+,20+,21+,22-,23+,24+/m0/s1
Formula Weight 461.3353
Chemical Name or Material Glucosyl-C18-sphingosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.