missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucose Transporter GLUT8 Antibody (VB1-050) [Janelia Fluor™ 585], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28056JF585
This item is not returnable.
View return policy
Description
Glucose Transporter GLUT8 Monoclonal antibody specifically detects Glucose Transporter GLUT8 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Glucose Transporter GLUT8 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| GLUT-8, GLUT8solute carrier family 2 (facilitated glucose transporter) member 8, GLUTX1facilitated glucose transporter member 8, solute carrier family 2 (facilitated glucose transporter), member 8 | |
| SLC2A8 | |
| 0.1 mL | |
| Alzheimers Research, Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 29988 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| VB1-050 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction