missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Glucose Transporter GLUT8 Antibody (VB1-050) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28056-1mg
This item is not returnable.
View return policy
Description
Glucose Transporter GLUT8 Monoclonal antibody specifically detects Glucose Transporter GLUT8 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Glucose Transporter GLUT8 | |
| Monoclonal | |
| Unconjugated | |
| Q9NY64 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| VB1-050 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| GLUT-8, GLUT8solute carrier family 2 (facilitated glucose transporter) member 8, GLUTX1facilitated glucose transporter member 8, solute carrier family 2 (facilitated glucose transporter), member 8 | |
| SLC2A8 | |
| 1 mg | |
| Alzheimers Research, Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 29988 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction