missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucose Transporter GLUT6 Antibody (OTI7E3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47987
This item is not returnable.
View return policy
Description
Glucose Transporter GLUT6 Monoclonal specifically detects Glucose Transporter GLUT6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Glucose Transporter GLUT6 | |
| Monoclonal | |
| 1.47 mg/mL | |
| Western Blot 1:200, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50 | |
| SLC2A6 | |
| Full-length protein expressed in 293T cell transfected with human SLC2A6 expression vector | |
| Affinity Purified | |
| RUO | |
| 11182 | |
| Human | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI7E3 | |
| Unconjugated | |
| Glucose transporter type 6, Glucose transporter type 9, GLUT6, GLUT-9, GLUT9GLUT-6, HSA011372, solute carrier family 2 (facilitated glucose transporter), member 6, solute carrier family 2, facilitated glucose transporter member 6 | |
| Mouse | |
| 54.5 kDa | |
| 0.1 mL | |
| Primary | |
| This antibody is specific for Homo sapiens solute carrier family 2 (facilitated glucose transporter), member 6 (SLC2A6), transcript variant 1. | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction