missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucose Monohydrate, LoGiCal
Description
Glucose Monohydrate
Specifications
Specifications
| Chemical Name or Material | Glucose Monohydrate |
| CAS | 14431-43-7 |
| Analyte or Component Names | Glucose Monohydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H12 O6 . H2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Glucose monohydrate, (+)-D-Glucopyranose monohydrate |
| IUPAC Name | (2 SR,3 R,4 S,5 S,6 R)-6-(hydroxymethyl)tetrahydropyran-2,3,4,5-tetrol monohydrate |
| Industry Type | Forensics |
| Molecular Weight (g/mol) | 198.17 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?