missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucose 6 phosphate isomerase, Mouse anti-Human, Clone: CPTC-GPI-1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79711-100UG
This item is not returnable.
View return policy
Description
Glucose 6 phosphate isomerase Monoclonal specifically detects Glucose 6 phosphate isomerase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Glucose 6 phosphate isomerase | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1-2 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| P06744 | |
| GPI | |
| Recombinant human full-length Glucose 6 phosphate isomerase protein | |
| 100 μg | |
| Cytokine Research, Phospho Specific | |
| 2821 | |
| Store at 4°C. | |
| IgG2a κ |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array | |
| CPTC-GPI-1 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| AMFGNPI, Autocrine motility factor, DKFZp686C13233, EC 5.3.1.9, glucose phosphate isomerase, glucose-6-phosphate isomerase, hexose monophosphate isomerase, hexosephosphate isomerase, neuroleukin, NLKSA36, oxoisomerase, PGI, PHI, Phosphoglucose isomerase, phosphohexomutase, Phosphohexose isomerase, phosphosaccharomutase, SA-36, Sperm antigen 36, sperm antigen-36 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction