missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Glucose 6 phosphate isomerase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29054
This item is not returnable.
View return policy
Description
Glucose 6 phosphate isomerase Polyclonal antibody specifically detects Glucose 6 phosphate isomerase in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| Glucose 6 phosphate isomerase | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| AMFGNPI, Autocrine motility factor, DKFZp686C13233, EC 5.3.1.9, glucose phosphate isomerase, glucose-6-phosphate isomerase, hexose monophosphate isomerase, hexosephosphate isomerase, neuroleukin, NLKSA36, oxoisomerase, PGI, PHI, Phosphoglucose isomerase, phosphohexomutase, Phosphohexose isomerase, phosphosaccharomutase, SA-36, Sperm antigen 36, sperm antigen-36 | |
| Between 1 and 50 | |
| 100 μg | |
| Cytokine Research, Phospho Specific | |
| 2821 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction