missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucose 6 phosphate isomerase Antibody (GPI/2439), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44601-20ug
This item is not returnable.
View return policy
Description
Glucose 6 phosphate isomerase Monoclonal antibody specifically detects Glucose 6 phosphate isomerase in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| Glucose 6 phosphate isomerase | |
| Monoclonal | |
| 0.2 mg/ml | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot | |
| GPI/2439 | |
| Unconjugated | |
| AMFGNPI, Autocrine motility factor, DKFZp686C13233, EC 5.3.1.9, glucose phosphate isomerase, glucose-6-phosphate isomerase, hexose monophosphate isomerase, hexosephosphate isomerase, neuroleukin, NLKSA36, oxoisomerase, PGI, PHI, Phosphoglucose isomerase, phosphohexomutase, Phosphohexose isomerase, phosphosaccharomutase, SA-36, Sperm antigen 36, sperm antigen-36 | |
| Recombinant human full-length Glucose 6 phosphate isomerase protein (Uniprot: P06744). | |
| 20 μg | |
| Cytokine Research, Phospho Specific | |
| 2821 | |
| Store at 4⪚C. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction