missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucose 6 Phosphate Dehydrogenase Rabbit anti-Human, Mouse, Rat, Clone: S02-6B6, Novus Biologicals™
Rabbit Monoclonal Antibody
£232.00 - £451.00
Specifications
| Antigen | Glucose 6 Phosphate Dehydrogenase |
|---|---|
| Clone | S02-6B6 |
| Dilution | Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18369871
|
Bio-Techne
NBP3-14935-25UL |
25 μg |
£232.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18325764
|
Novus Biologicals
NBP3-14935-100UL |
100 μg |
£451.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glucose 6 Phosphate Dehydrogenase Monoclonal antibody specifically detects Glucose 6 Phosphate Dehydrogenase in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Glucose 6 Phosphate Dehydrogenase | |
| Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| G6PD, G6PD1EC 1.1.1.49, glucose-6-phosphate dehydrogenase | |
| Recombinant protein of human Glucose 6 Phosphate Dehydrogenase (Uniprot#: P11413) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S02-6B6 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cellular Markers, Golgi Apparatus Markers | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| 2539 | |
| IgG | |
| Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title