missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucose 6 Phosphate Dehydrogenase Rabbit anti-Human, Clone: 6V8O9, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Glucose 6 Phosphate Dehydrogenase Monoclonal antibody specifically detects Glucose 6 Phosphate Dehydrogenase in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Glucose 6 Phosphate Dehydrogenase |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 6V8O9 |
| Conjugate | Unconjugated |
| Formulation | PBS, 0.05% BSA, 50% glycerol, pH7.3 |
| Gene Alias | G6PD, G6PD1EC 1.1.1.49, glucose-6-phosphate dehydrogenase |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 400-500 of human Glucose 6 Phosphate Dehydrogenase (P11413). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?