missing translation for 'onlineSavingsMsg'
Learn More

Glucose 6 Phosphate Dehydrogenase Antibody (2F6), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18405514 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405514 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405514 Supplier Novus Biologicals Supplier No. NBP130358MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Glucose 6 Phosphate Dehydrogenase Monoclonal antibody specifically detects Glucose 6 Phosphate Dehydrogenase in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Isotype IgG2b κ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.