missing translation for 'onlineSavingsMsg'
Learn More

Glucose 6 Phosphate Dehydrogenase Antibody (2F6), DyLight 488, Novus Biologicals™

Product Code. 18415224 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415224 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415224 Supplier Novus Biologicals Supplier No. NBP130358G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Glucose 6 Phosphate Dehydrogenase Monoclonal antibody specifically detects Glucose 6 Phosphate Dehydrogenase in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glucose 6 Phosphate Dehydrogenase
Applications Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 2F6
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Accession No. NP_001035810.1
Gene Alias G6PD, G6PD1EC 1.1.1.49, glucose-6-phosphate dehydrogenase
Host Species Mouse
Immunogen Recombinant human Glucose 6 Phosphate Dehydrogenase (35-506aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 2539
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.