missing translation for 'onlineSavingsMsg'
Learn More

Glucosamine (N-acetyl)-6-Sulfatase/GNS Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18334422 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18334422 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18334422 Supplier Novus Biologicals Supplier No. NBP299216AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Glucosamine (N-acetyl)-6-Sulfatase/GNS Polyclonal specifically detects Glucosamine (N-acetyl)-6-Sulfatase/GNS in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Glucosamine (N-acetyl)-6-Sulfatase/GNS
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 3.1.6, EC 3.1.6.14, G6Sglucosamine-6-sulfatase, glucosamine (N-acetyl)-6-sulfatase, Glucosamine-6-sulfatase, MGC21274, N-acetylglucosamine-6-sulfatase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Glucosamine (N-acetyl)-6-Sulfatase/GNS (NP_002067.1; Met1-Leu552).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2799
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.