missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucosamine (N-acetyl)-6-Sulfatase/GNS Antibody (007), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89413APC
This item is not returnable.
View return policy
Description
Glucosamine (N-acetyl)-6-Sulfatase/GNS Monoclonal antibody specifically detects Glucosamine (N-acetyl)-6-Sulfatase/GNS in Human samples. It is validated for Western Blot, ELISA
Specifications
| Glucosamine (N-acetyl)-6-Sulfatase/GNS | |
| Monoclonal | |
| APC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 2799 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| 007 | |
| Western Blot, ELISA | |
| EC 3.1.6, EC 3.1.6.14, G6Sglucosamine-6-sulfatase, glucosamine (N-acetyl)-6-sulfatase, Glucosamine-6-sulfatase, MGC21274, N-acetylglucosamine-6-sulfatase | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Glucosamine (N-acetyl)-6-Sulfatase/GNS (Accession#: NP_002067.1; Met 1-Leu 552). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction