missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucoraphanin Potassium Salt (Contains up to ~15% Inorganics), TRC
Specifications
Specifications
| Chemical Name or Material | Glucoraphanin potassium salt |
| Analyte or Component Names | Glucoraphanin potassium salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H22 N O10 S3 . K |
| Synonym | Sulforaphane glucosinolate potassium salt, 4-Methylsulfinylbutyl glucosinolate potassium salt |
| InChI Formula | InChI=1S/C12H23NO10S3.K/c1-25(18)5-3-2-4-8(13-23-26(19,20)21)24-12-11(17)10(16)9(15)7(6-14)22-12;/h7,9-12,14-17H,2-6H2,1H3,(H,19,20,21);/q;+1/p-1/t7-,9-,10+,11-,12+,25?;/m1./s1 |
| Solution Type | Neat |
| SMILES | [K+].CS(=O)CCCCC(=NOS(=O)(=O)[O-])S[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O |
| IUPAC Name | potassium;[[5-methylsulfinyl-1-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydropyran-2-yl]sulfanyl-pentylidene]amino] sulfate |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?