missing translation for 'onlineSavingsMsg'
Learn More

Glucoraphanin Potassium Salt (Contains up to ~15% Inorganics), TRC

Product Code. 30402127
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
Unit Size:
1mg
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402127 2.5 mg 2.5mg
30396576 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402127 Supplier Toronto Research Chemicals Supplier No. TRCG4502552.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glucoraphanin potassium salt
Analyte or Component Names Glucoraphanin potassium salt
Shipping Condition Room Temperature
Molecular Formula C12 H22 N O10 S3 . K
Synonym Sulforaphane glucosinolate potassium salt, 4-Methylsulfinylbutyl glucosinolate potassium salt
InChI Formula InChI=1S/C12H23NO10S3.K/c1-25(18)5-3-2-4-8(13-23-26(19,20)21)24-12-11(17)10(16)9(15)7(6-14)22-12;/h7,9-12,14-17H,2-6H2,1H3,(H,19,20,21);/q;+1/p-1/t7-,9-,10+,11-,12+,25?;/m1./s1
Solution Type Neat
SMILES [K+].CS(=O)CCCCC(=NOS(=O)(=O)[O-])S[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
IUPAC Name potassium;[[5-methylsulfinyl-1-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydropyran-2-yl]sulfanyl-pentylidene]amino] sulfate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 475.6
Quantity 2.5 mg
Formula Weight 475.0043
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.