missing translation for 'onlineSavingsMsg'
Learn More

Gluconate sodium, MedChemExpress

Product Code. 16438606
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438606 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16438606 Supplier Medchem Express Supplier No. HYB1092A
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Gluconate sodium (D-Gluconic acid sodium salt) is a corrosion and scale inhibitor of ordinary steel in simulated cooling water.

Gluconate sodium (D-Gluconic acid sodium salt) is a corrosion and scale inhibitor of ordinary steel in simulated cooling water.

Specifications

Chemical Name or Material Gluconate sodium
CAS 527-07-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C6H11NaO7
Quantity 100 mg
Synonym D-Gluconic acid sodium salt Sodium D-gluconate D-Gluconate sodium salt
Solubility Information H2O : ≥ 100 mg/mL (458.42 mM) ∣DMSO : 1 mg/mL (4.58 mM; ultrasonic and warming and heat to 60°C)
SMILES O[C@H]([C@H]([C@@H]([C@@H](CO)O)O)O)C(O[Na])=O
Molecular Weight (g/mol) 218.14
Formula Weight 218.14
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.