missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucokinase/GCK Antibody (OTI3E3), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70822AF488
This item is not returnable.
View return policy
Description
Glucokinase/GCK Monoclonal specifically detects Glucokinase/GCK in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Glucokinase/GCK | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GCK | |
| Full length recombinant protein of human Glucokinase/GCK (NP_000153) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI3E3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| EC 2.7.1, EC 2.7.1.2, FGQTL3, GK, GLK, glucokinase (hexokinase 4), hexokinase-4, Hexokinase-D, HK4pancreatic isozyme, HKIV, HXKP, LGLK, maturity onset diabetes of the young 2, MODY2 | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism, Protein Kinase | |
| 2645.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction