missing translation for 'onlineSavingsMsg'
Learn More

Glucokinase/GCK Antibody (223), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18323211 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18323211 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18323211 Supplier Novus Biologicals Supplier No. NBP289787AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Glucokinase/GCK Monoclonal specifically detects Glucokinase/GCK in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glucokinase/GCK
Applications Western Blot, ELISA
Classification Monoclonal
Clone 223
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias EC 2.7.1, EC 2.7.1.2, FGQTL3, GK, GLK, glucokinase (hexokinase 4), hexokinase-4, Hexokinase-D, HK4pancreatic isozyme, HKIV, HXKP, LGLK, maturity onset diabetes of the young 2, MODY2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Glucokinase/GCK (NP_000153.1; Leu2-Gln465).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2645
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.