missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Glucocorticoid Receptor, Ligand Binding Domain, GST Tag Recombinant Protein

Product Code. 15410714
Change view
Click to view available options
Quantity:
4.5 mg
Unit Size:
4.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15410714 4.5 mg 4.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15410714 Supplier Invitrogen™ Supplier No. A15668

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Glucocorticoid receptor ligand binding domain (GR-LBD) is a 57.85 kDa recombinant human protein (amino acids 521-777) expressed as a glutathione-S-transferast (GST) fusion protein in baculovirus-infected insect cells. GR-LBD is expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator displacement studies. GR-LBD human recombinant protein is: , Flexible—use in your in-house assay or as part of our LanthaScreen™ Coactivator or Competitive Binding assays for a complete assay solution, Ready to use—active and tagged (with GST), Convenient—available in bulk sizes Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond. Protein Form: Ligand Binding Domain, Recombinant

Glucocorticoid Receptor (NR3C1) is a receptor for glucocorticoids that can act as both a transcription factor and as a regulator of other transcription factors. Glucocorticoid Receptor can also be found in heteromeric cytoplasmic complexes along with heat shock factors and immunophilins. The protein is typically found in the cytoplasm until it binds a ligand, which induces transport into the nucleus. Glucocorticoid Receptor is expressed in the heart, detected in left and right atria, left and right ventricles, aorta, apex, intraventricular septum, and atrioventricular node as well as whole adult and fetal heart. Alternate splicing, the use of at least three different promoters, and alternate translation initiation sites result in several transcript variants encoding the same protein or different isoforms, but the full-length nature of some variants has not been determined. Mutations in the Glucocorticoid Receptor gene are a cause of glucocorticoid resistance, or cortisol, resistance. Studies have shown that glucocorticoid receptor (GR) must be associated with a complex of chaperone proteins for ligand activation. GR binds to known steroids such as dexamethasone with nanomolar affinity
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P04150
Concentration See Label
For Use With (Application) Kinase Assay
Formulation proprietary buffer with no preservative
Gene ID (Entrez) 2908
Name Human Glucocorticoid Receptor, Ligand Binding Domain, GST Tag
Quantity 4.5 mg
Regulatory Status RUO - research use only
Gene Alias cytosolic/nuclear receptor; fb13f09; GCCR; Gcr; GCRST; glucocorticoid nuclear receptor variant 1; glucocorticoid receptor; glucocorticoid receptor 1; glucocorticoid receptor alpha; GR; gr alpha; GR Kit; Grl; Grl1; Grl-1; Nr3c1; Nuclear receptor subfamily 3 group C member 1; nuclear receptor subfamily 3, group C, member 1; nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor); OTTHUMP00000222854; OTTHUMP00000222858; utouto; utut; wu:fb13f09; zgc:113038
Common Name Glucocorticoid Receptor (Nr3c1)
Gene Symbol NR3C1
Protein Length 521-777
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Baculovirus
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.