Learn More
Invitrogen™ Glucocorticoid Receptor alpha Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA1516
Description
PA1-516 detects glucocorticoid receptor (GR) alpha from human, mouse and rat tissues. This antibody does not detect GR beta. PA1-516 has been successfully used in Western blot, immunocytochemistry, immunofluorescence and immunoprecipitation (ChIP) procedures. By Western blot, this antibody detects an ~94 kDa protein representing GR in HeLa cell lysate. PA1-516 immunizing peptide corresponds to amino acid residues 755-771 of human GR alpha. This peptide (Cat. # PEP-221) is available for use in neutralization and control experiments. Reconstitute with 50 μL of distilled water.
Glucocorticoid Receptor (NR3C1) is a receptor for glucocorticoids that can act as both a transcription factor and as a regulator of other transcription factors. Glucocorticoid Receptor can also be found in heteromeric cytoplasmic complexes along with heat shock factors and immunophilins. The protein is typically found in the cytoplasm until it binds a ligand, which induces transport into the nucleus. Glucocorticoid Receptor is expressed in the heart, detected in left and right atria, left and right ventricles, aorta, apex, intraventricular septum, and atrioventricular node as well as whole adult and fetal heart. Alternate splicing, the use of at least three different promoters, and alternate translation initiation sites result in several transcript variants encoding the same protein or different isoforms, but the full-length nature of some variants has not been determined. Mutations in the Glucocorticoid Receptor gene are a cause of glucocorticoid resistance, or cortisol, resistance. Studies have shown that glucocorticoid receptor (GR) must be associated with a complex of chaperone proteins for ligand activation. GR binds to known steroids such as dexamethasone with nanomolar affinity.
Specifications
| Glucocorticoid Receptor alpha | |
| Polyclonal | |
| Unconjugated | |
| Nr3c1 | |
| cytosolic/nuclear receptor; fb13f09; GCCR; Gcr; GCRST; glucocorticoid nuclear receptor variant 1; glucocorticoid receptor; glucocorticoid receptor 1; glucocorticoid receptor alpha; GR; gr alpha; GR Kit; Grl; Grl1; Grl-1; Nr3c1; Nuclear receptor subfamily 3 group C member 1; nuclear receptor subfamily 3, group C, member 1; nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor); OTTHUMP00000222854; OTTHUMP00000222858; utouto; utut; wu:fb13f09; zgc:113038 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14815, 24413, 2908 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P04150, P06536, P06537 | |
| Nr3c1 | |
| Synthetic Peptide: C(755) E I I T N Q I P K Y S N G N I K K(771). | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
- Glucocorticoid Receptor alpha Antibody
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H302-Harmful if swallowed.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P330-Rinse mouth.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : SODIUM AZIDE
Your input is important to us. Please complete this form to provide feedback related to the content on this product.