missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucoamylase Antibody [Janelia Fluor™ 635], Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-78076JF635
This item is not returnable.
View return policy
Description
Glucoamylase Polyclonal antibody specifically detects Glucoamylase in Fungi samples. It is validated for ELISA,Western Blot
Specifications
| Glucoamylase | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Glucoamylase [Rhizopus] (Uniprot: P07683) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Western Blot | |
| Janelia Fluor 635 | |
| Goat | |
| Multi-step | |
| RUO | |
| Fungi | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction