missing translation for 'onlineSavingsMsg'
Learn More

Glucagon R/GCGR, Mouse anti-Human, Clone: hGR-2 F6, Novus Biologicals™

Mouse Monoclonal Antibody

£207.00 - £463.00

Specifications

Antigen Glucagon R/GCGR
Clone hGR-2 F6
Concentration 1 mg/mL
Dilution Immunocytochemistry/Immunofluorescence, Block/Neutralize
Applications Immunofluorescence, Blocking Assay
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18387890
View Documents
Novus Biologicals
NBP2-81065-0.025MG
0.025 mg
£207.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18339570
View Documents
Novus Biologicals
NBP2-81065-0.2MG
0.2 mg
£463.00
0.20mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Glucagon R/GCGR Monoclonal specifically detects Glucagon R/GCGR in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Block/Neutralize.
Specifications

Specifications

Glucagon R/GCGR
1 mg/mL
Immunofluorescence, Blocking Assay
Unconjugated
Mouse
GPCR, Signal Transduction
PBS
2642
Recombinant human Glucagon R/GCGR.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
hGR-2 F6
Immunocytochemistry/Immunofluorescence, Block/Neutralize
Monoclonal
Purified
RUO
Human
FLJ97182, GGR, GL-R, glucagon receptor, MGC138246
GCGR
IgG1 κ
Protein A purified
The antibody binds to the human glucagon receptor, Glucagon R/GCGR.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.