missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucagon R/GCGR Antibody (hGR-2 F6), mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Glucagon R/GCGR Monoclonal antibody specifically detects Glucagon R/GCGR in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Neutralization
Specifications
Specifications
| Antigen | Glucagon R/GCGR |
| Conjugate | mFluor Violet 450 SE |
| Gene Alias | FLJ97182, GGR, GL-R, glucagon receptor, MGC138246 |
| Host Species | Mouse |
| Gene ID (Entrez) | 2642 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?